Learn More
Research Products International Corp Yeast Extract, Granulated, 5 Kilograms
SDP
Catalog No. 50489153
Encompass_Preferred
Dried yeast autolysate with a high content of amino nitrogen and water soluble B complex vitamins. Material is specially granulated to reduce dust and increase solubility rate.
Suitable for Microbiology Applications.
Catalog No. 50-489-153 Supplier Research Products International Corp Supplier No. Y200255000.0
Dried yeast autolysate with a high content of amino nitrogen and water soluble B complex vitamins. Material is specially granulated to reduce dust and increase solubility rate.
Suitable for Microbiology Applications.