Learn More
HIMEDIA Yeast Extract, Granulated, RMG027, Microbiology Culture Media Ingredients, Microbial Growth Supplement, Nitrogen and Vitamin Source, Low Salt, Free Flowing, Granules, 25 kg
Catalog No. NC4013790
Encompass_Preferred
Yeast Extract, Granulated is a rich source of amino acids, B-group vitamins and growth factors prepared from a selected strain of Saccharomyces under controlled conditions. The granulated form flows freely and resists lump formation, dissolving cleanly during media preparation, while its low salt content suits a wide range of formulations. It is widely used to enrich microbiological culture media and to support the mass cultivation of bacteria, yeasts and other micro-organisms in research and production. Store in a cool dry place. Packaged as 25 kg.
Catalog No. NC4013790 Supplier HIMEDIA Supplier No. RMG02725KG
Yeast Extract, Granulated is a rich source of amino acids, B-group vitamins and growth factors prepared from a selected strain of Saccharomyces under controlled conditions. The granulated form flows freely and resists lump formation, dissolving cleanly during media preparation, while its low salt content suits a wide range of formulations. It is widely used to enrich microbiological culture media and to support the mass cultivation of bacteria, yeasts and other micro-organisms in research and production. Store in a cool dry place. Packaged as 25 kg.