Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yeast Extract, Powder, MP Biomedicals™

Catalog No. MP219402783 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219402783 250 g
ICN19402791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219402783 Supplier MP Biomedicals, Inc Supplier No. 0219402783
Add to Cart
Add to Cart

Yeast extract From Yeast, Molecular Biology Reagent, Yellow-tan Powder

  • It is used as a bacteriological media component for a variety of microorganisms and molecular genetics applications.
  • Yeast extract powder is a vacuum dried autolyzed yeast extract produced from 100% Baker's yeast.
  • It is a concentrated source of soluble protein components, carbohydrates, and B Complex vitamins natural to yeast.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8013-01-2

Specifications

Color Yellow-tan
Content And Storage Store at Room Temperature (15-30°C). Store Desiccated.
Form Powder
Purity or Quality Grade Molecular Biology Grade
Quantity 250 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.