Learn More
Crescent Chemical Co Inc YEAST EXTRAXCT SERVABACTER
SDP
Supplier:  Crescent Chemical Co Inc SE24540.02
Encompass_Preferred
Yeast extract SERVABACTER powder Water soluble fraction of yeast autolysate. A 2 % aqueous solution is clear and has a pH of ca. 6.5 - 7.5. Rich in B-vitamins and growth factors, convenient standard material for culture media. Tested for use in tissue culture. Size -
Catalog No. 50-247-711