Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Yeast Hydrolysate Enzymatic, MP Biomedicals™

Catalog No. MP210330480 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210330480 100 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210330480 Supplier MP Biomedicals™ Supplier No. 0210330480
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

an autolyzed yeast extract that is a concentrated source of soluble protein components, carbohydrates and B-complex vitamins that are natural to yeast.

  • Produced from 100% brewer’s yeast.
  • Brown-colored powder, soluble in water
  • Store at room temperature
TRUSTED_SUSTAINABILITY

Specifications

Color Brown
Description Yeast hydrolysate enzymatic
Content And Storage Store at Room Temperature
Form Powder
For Use With (Application) Source of soluble protein components; carbohydrates; and B Complex vitamins
Quantity 100 lb.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.