Learn More
Description
- Soluble in water
Specifications
Specifications
| Description | Enzymatic |
| Form | Light tan powder |
| Product Type | Yeast |
| Format | Plastic bottle |
| pH | 5.4 to 6.2 |
| Purity or Quality Grade | Ultrapure |
| Quantity | 50 lb. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.