Learn More
Description
- 0379P Candida albicans derived from NCYC 1363
- 0992P Candida glabrata derived from ATCC™ 2001™
- 0726P Candida parapsilosis derived from ATCC™ 22019™
- 01051P Cryptococcus gattii derived from ATCC™ MYA-4560™
- 0736P Saccharomyces cerevisiae derived from NCYC 79
Specifications
Specifications
| Format | KWIK-STIK |
| Manufacturing Origin | United States |
| Product Type | QC Set |
| Sterility | Sterile |
| Target Organism Class | Fungi |
| Test Type | Microbial Identification, Infectious Disease, Fungal Testing, Qualitative QC |
| Includes | 2 each: Candida albicans, Candida glabrata, Candida parapsilosis, Cryptococcus gattii, Saccharomyces cerevisiae |
| Quantity | 10 KWIK-STIKs |
| Regulatory Status | IVD |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
