Learn More
Bioworld Yeast Lysis Solution, 500 mL
Catalog No. 50198467
Encompass_Preferred
Yeast Lysis Solution-DNA Isolation Common solution for protein and DNA isolation from yeast. Technical Information:LiCl (2.5M)Tris-HCl, pH 8.0 (50mM)Triton X 100 (1%)EDTA (62.5mM)
Catalog No. 50-198-467 Supplier Bioworld Supplier No. 208400232
Yeast Lysis Solution-DNA Isolation Common solution for protein and DNA isolation from yeast. Technical Information:LiCl (2.5M)Tris-HCl, pH 8.0 (50mM)Triton X 100 (1%)EDTA (62.5mM)