Learn More
MOLTOX Molecular Toxicology Inc Yeast Malt Agar (low pH), 20 mL/plate, 20 plates/sleeve, Store at Room Temperature
SDP
Catalog No. 50948697
Encompass_Preferred
Yeast Malt Agar, or Yeast Mold Agar, is used for the cultivation of yeasts, molds and other acidophic microorganisms. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-697 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S88
Yeast Malt Agar, or Yeast Mold Agar, is used for the cultivation of yeasts, molds and other acidophic microorganisms. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.