Learn More
Takara Bio YEAST MEDIA TRP DO SUPPLMNT
SDP
Supplier:  Takara Bio 630413
Encompass
Minimal SD Agar Basea, 467g, Comprised of a yeast nitrogen base, ammonium sulfate, and a carbon source, Ideal for use with Matchmaker One-hybrid and Two-hybrid systems
Catalog No. NC9413248