Learn More
New England Biolabs, Inc. Yeast Medium Pack – 12 g
SDP
Catalog No. 50994921
Encompass
The Yeast Carbon Base Medium Powder contains sufficient reagents needed to make 1 liter of Yeast Carbon Base (YCB) Agar Medium containing 5 mM acetamide. YCB Agar Medium is used for acetamide selection of K. lactis cells that have been transformed with a pKLAC1-based expression vector (NEB #N3740).
Catalog No. 50-994-921 Supplier New England Biolabs, Inc. Supplier No. B9017S
The Yeast Carbon Base Medium Powder contains sufficient reagents needed to make 1 liter of Yeast Carbon Base (YCB) Agar Medium containing 5 mM acetamide. YCB Agar Medium is used for acetamide selection of K. lactis cells that have been transformed with a pKLAC1-based expression vector (NEB #N3740).