Learn More
Alkali Scientific Yeast Mold (YM) Agar, 15x100mm, 25mL, 10/pk
SDP
Catalog No. 5000662586
Encompass_Preferred
Yeast Mold (YM) Agar, 15×100mm plates containing 25 mL of medium, is a general purpose culture medium formulated for the cultivation, isolation, and enumeration of yeasts and molds from clinical, environmental, and food related samples. The nutrient rich formulation supports robust fungal growth while allowing researchers to observe colony morphology and other characteristics useful for identification. YM agar is commonly used in mycology research, environmental monitoring, and quality control laboratories for the recovery and maintenance of fungal organisms. The balanced nutrient composition supports both yeast and mold development while maintaining reliable growth performance. Each plate is pre poured with sterile agar to a consistent depth, ensuring uniform media distribution, dependable growth conditions, and reproducible laboratory results.
Catalog No. 50-006-62586 Supplier Alkali Scientific Supplier No. YMAP00
Yeast Mold (YM) Agar, 15×100mm plates containing 25 mL of medium, is a general purpose culture medium formulated for the cultivation, isolation, and enumeration of yeasts and molds from clinical, environmental, and food related samples. The nutrient rich formulation supports robust fungal growth while allowing researchers to observe colony morphology and other characteristics useful for identification. YM agar is commonly used in mycology research, environmental monitoring, and quality control laboratories for the recovery and maintenance of fungal organisms. The balanced nutrient composition supports both yeast and mold development while maintaining reliable growth performance. Each plate is pre poured with sterile agar to a consistent depth, ensuring uniform media distribution, dependable growth conditions, and reproducible laboratory results.