Learn More
Bioworld Yeast Nitrogen Base, 250 g
Catalog No. 50254397
Encompass_Preferred
Yeast Nitrogen BaseFor classifying yeasts based on carbon assimilationRequires addition of a carbon sourcepH: 5.4Working Concentration: 6.7 g/L (single-strength) or 67 g/L (10X)Dehydrated Appearance: off-white, homogeneous powderPrepared Appearance: single-strength solution is colorless to very pale yellow agar; 10X solution is yellow agarSoluble in waterContains: Ammonium sulfateMonopotassium phosphateMagnessium sulfateCalcium chlorideSodium ChlorideHistidine HCIMethionineTryptophanInositolThis product is sold for laboratory research use only.
Catalog No. 50-254-397 Supplier Bioworld Supplier No. 306201001
Yeast Nitrogen BaseFor classifying yeasts based on carbon assimilationRequires addition of a carbon sourcepH: 5.4Working Concentration: 6.7 g/L (single-strength) or 67 g/L (10X)Dehydrated Appearance: off-white, homogeneous powderPrepared Appearance: single-strength solution is colorless to very pale yellow agar; 10X solution is yellow agarSoluble in waterContains: Ammonium sulfateMonopotassium phosphateMagnessium sulfateCalcium chlorideSodium ChlorideHistidine HCIMethionineTryptophanInositolThis product is sold for laboratory research use only.