Learn More
Research Products International Corp Yeast Nitrogen Base without Amino Acids and Ammonium Sulfate, 250 Grams
SDP
Catalog No. 50489163
Encompass_Preferred
For classifying yeasts based on amino acid and carbohydrate requirements. Contains all essential vitamins and inorganic salts necessary for the cultivation of yeasts. Excludes histidine, methionine, tryptophan, carbon and nitrogen source. Also Contains <1 mg/l each of vitamins and trace inorganic salts.
Catalog No. 50-489-163 Supplier Research Products International Corp Supplier No. Y20060250.0
For classifying yeasts based on amino acid and carbohydrate requirements. Contains all essential vitamins and inorganic salts necessary for the cultivation of yeasts. Excludes histidine, methionine, tryptophan, carbon and nitrogen source. Also Contains <1 mg/l each of vitamins and trace inorganic salts.