Learn More
Research Products International Corp Yeast Nitrogen Base without Amino Acids, 250 Grams
SDP
Catalog No. 50489160
Encompass_Preferred
For classifying yeasts based on amino acid and carbohydrates requirements. Contains all essential vitamins and inorganic salts necessary for the cultivaiton of yeasts. Excludes histidine methionine, tryptophan and carbon source. Also contains <1 mg/L each of vitamins and trace inorganic salts.
Catalog No. 50-489-160 Supplier Research Products International Corp Supplier No. Y20040250.0
For classifying yeasts based on amino acid and carbohydrates requirements. Contains all essential vitamins and inorganic salts necessary for the cultivaiton of yeasts. Excludes histidine methionine, tryptophan and carbon source. Also contains <1 mg/L each of vitamins and trace inorganic salts.