Learn More
Research Products International Corp Yeast Nitrogen Base with Amino Acids, 250 Grams
SDP
Catalog No. 50489156
Encompass_Preferred
For classifying yeasts based on amino acid and carbohydrates requirements. Contains all essential vitamins and inorganic salts necessary for the cultivation of yeasts. Excludes carbohydrate source.
Catalog No. 50-489-156 Supplier Research Products International Corp Supplier No. Y20030250.0
For classifying yeasts based on amino acid and carbohydrates requirements. Contains all essential vitamins and inorganic salts necessary for the cultivation of yeasts. Excludes carbohydrate source.