Learn More
Sigma Aldrich Fine Chemicals Biosciences Yeast Nitrogen Base With A250G
Catalog No. 502904153
Encompass
Yeast Nitrogen Base is a highly-referenced growth medium used for the cultivation of yeast. This nutrient-rich microbial broth contains amino acids nitrogen vitamins trace elements and salts.
Catalog No. 50-290-4153 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Y1250250G
Yeast Nitrogen Base is a highly-referenced growth medium used for the cultivation of yeast. This nutrient-rich microbial broth contains amino acids nitrogen vitamins trace elements and salts.