Learn More
Sigma Aldrich Fine Chemicals Biosciences Yeast Nitrogen Base Withou250G
Catalog No. 502901647
Encompass
Yeast Nitrogen Base is considered ideal for culturing wild type yeast. The presence of amino acid is known to prevent selectable marker utilization. Hence this media is formulated without amino acids and is useful for cloning and manipulation of yeast artificial chromosome.
Catalog No. 50-290-1647 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Y0626250G
Yeast Nitrogen Base is considered ideal for culturing wild type yeast. The presence of amino acid is known to prevent selectable marker utilization. Hence this media is formulated without amino acids and is useful for cloning and manipulation of yeast artificial chromosome.