Learn More
Alkali Scientific Yeast Nitrogen Base without Amino Acids, 1 Kg
SDP
Catalog No. 5000661524
Encompass_Preferred
Yeast Nitrogen Base without Amino Acids, 1Kg, is a versatile nutrient base used to support the growth of yeasts, molds, and other fungi in laboratory research. Unlike typical yeast nitrogen bases, this product does not contain amino acids, making it suitable for experiments where specific amino acids need to be added separately. The 1kg size is ideal for high-volume laboratories or those involved in large-scale research projects requiring consistent media preparation for fungal cultures, yeast propagation, and fermentation studies. It is commonly used in microbiological and biochemical applications where precise nutrient control is critical for reliable results.
Catalog No. 50-006-61524 Supplier Alkali Scientific Supplier No. AST002CP
Yeast Nitrogen Base without Amino Acids, 1Kg, is a versatile nutrient base used to support the growth of yeasts, molds, and other fungi in laboratory research. Unlike typical yeast nitrogen bases, this product does not contain amino acids, making it suitable for experiments where specific amino acids need to be added separately. The 1kg size is ideal for high-volume laboratories or those involved in large-scale research projects requiring consistent media preparation for fungal cultures, yeast propagation, and fermentation studies. It is commonly used in microbiological and biochemical applications where precise nutrient control is critical for reliable results.