Learn More
Alkali Scientific Yeast Nitrogen Base without Amino Acids, 5 Kg
SDP
Catalog No. 5000661072
Encompass_Preferred
Yeast Nitrogen Base without Amino Acids is a high-quality, sterile powder used in microbiological research. It provides essential nutrients for yeast growth, excluding amino acids, making it ideal for controlled experimental conditions where amino acid supplementation is not required. This 5 kg powder is typically used in yeast propagation, fermentation studies, and metabolic experiments where a nitrogen base is needed. The absence of amino acids allows researchers to explore specific pathways of nitrogen metabolism or study the yeast's ability to synthesize amino acids under specific conditions. This medium is suitable for industrial and laboratory-scale studies requiring precise and reproducible results.
Catalog No. 50-006-61072 Supplier Alkali Scientific Supplier No. AST005CP
Yeast Nitrogen Base without Amino Acids is a high-quality, sterile powder used in microbiological research. It provides essential nutrients for yeast growth, excluding amino acids, making it ideal for controlled experimental conditions where amino acid supplementation is not required. This 5 kg powder is typically used in yeast propagation, fermentation studies, and metabolic experiments where a nitrogen base is needed. The absence of amino acids allows researchers to explore specific pathways of nitrogen metabolism or study the yeast's ability to synthesize amino acids under specific conditions. This medium is suitable for industrial and laboratory-scale studies requiring precise and reproducible results.