Learn More
Alkali Scientific Yeast Nitrogen Base without Amino Acids and Ammonium Sulfate, 1 Kg
SDP
Catalog No. 5000660875
Encompass_Preferred
Yeast Nitrogen Base without Amino Acids and Ammonium Sulfate is a specialized nutrient medium designed for yeast and fungal research. In this 1 kg powdered form, it is used in microbiological applications that require a controlled nitrogen source without additional amino acids or ammonium sulfate. This allows researchers to study yeast metabolism under conditions where nitrogen is the only added nutrient. The absence of ammonium sulfate provides a more specific approach to testing nitrogen sources. The medium is ideal for metabolic studies, fermentation processes, and applications that focus on nitrogen utilization by yeasts, allowing for enhanced experimental control and precision in biological assays.
Catalog No. 50-006-60875 Supplier Alkali Scientific Supplier No. STW001CP
Yeast Nitrogen Base without Amino Acids and Ammonium Sulfate is a specialized nutrient medium designed for yeast and fungal research. In this 1 kg powdered form, it is used in microbiological applications that require a controlled nitrogen source without additional amino acids or ammonium sulfate. This allows researchers to study yeast metabolism under conditions where nitrogen is the only added nutrient. The absence of ammonium sulfate provides a more specific approach to testing nitrogen sources. The medium is ideal for metabolic studies, fermentation processes, and applications that focus on nitrogen utilization by yeasts, allowing for enhanced experimental control and precision in biological assays.