Learn More
Bioworld Yeast Nitrogen Base (YNB), His Dropout, 1 L
Supplier:  Bioworld 306260111
Encompass_Preferred
Yeast Nitrogen Base (YNB)May be used to test carbon assimilation in the presence of a carbon source like glucose, galactose, or raffinoseBased upon the formulation of Yeast Agar except that Ammonium Sulfate is the sole nitrogen source and that addition of carbon source is requiredDoes not contain HistidinepH: 5.4Dehydrated Appearance: off-white, homogeneous powderPrepared Appearance: colorlessSoluble in water with agitationContains:Ammonium Sulfate LD-Methionine LD-Tryptophan Biotin Calcium PantothenateFolic Acid Inositol Niacinp-Aminobenzoic AcidPyridoxine HydrochlorideRiboflavinThiamine Hydrochloride Boric Acid Copper Sulfate Potassium Iodide Ferric Chloride Manganese Sulfate Sodium Molybdate Zinc Sulfate Monopotassium Phosphate Magnesium Sulfate Sodium Chloride Calcium Chloride
Catalog No. 50-254-483