Learn More
Bioworld Yeast Nitrogen Base (YNB), Trp Dropout, 1 L
Catalog No. 50254485
Encompass_Preferred
Yeast Nitrogen Base (YNB)May be used to test carbon assimilation in the presence of a carbon source like glucose, galactose, or raffinoseBased upon the formulation of Yeast Agar except that Ammonium Sulfate is the sole nitrogen source and that addition of carbon source is requiredDoes not contain TryptophanpH: 5.4Dehydrated Appearance: off-white, homogeneous powderPrepared Appearance: colorlessSoluble in water with agitationContains:Ammonium Sulfate L-Histidine MonohydrochlorideLD-Methionine Biotin Calcium PantothenateFolic Acid Inositol Niacinp-Aminobenzoic AcidPyridoxine HydrochlorideRiboflavinThiamine Hydrochloride Boric Acid Copper Sulfate Potassium Iodide Ferric Chloride Manganese Sulfate Sodium Molybdate Zinc Sulfate Monopotassium Phosphate Magnesium Sulfate Sodium Chloride Calcium Chloride
Catalog No. 50-254-485 Supplier Bioworld Supplier No. 306260131
Yeast Nitrogen Base (YNB)May be used to test carbon assimilation in the presence of a carbon source like glucose, galactose, or raffinoseBased upon the formulation of Yeast Agar except that Ammonium Sulfate is the sole nitrogen source and that addition of carbon source is requiredDoes not contain TryptophanpH: 5.4Dehydrated Appearance: off-white, homogeneous powderPrepared Appearance: colorlessSoluble in water with agitationContains:Ammonium Sulfate L-Histidine MonohydrochlorideLD-Methionine Biotin Calcium PantothenateFolic Acid Inositol Niacinp-Aminobenzoic AcidPyridoxine HydrochlorideRiboflavinThiamine Hydrochloride Boric Acid Copper Sulfate Potassium Iodide Ferric Chloride Manganese Sulfate Sodium Molybdate Zinc Sulfate Monopotassium Phosphate Magnesium Sulfate Sodium Chloride Calcium Chloride