Learn More
Zymo Research Corporation Yeast Protein Kit™, 200 preps
SDP
Catalog No. NC9993470
Encompass_Preferred
The kit essentially generates spheroplasts of yeast cells for subsequent analysis. It is optimized for S. cerevisiae and C. albicans. The kit can also be used for any fungus species that is susceptible to yeast lytic enzyme digestion.
Catalog No. NC9993470 Supplier Zymo Research Corporation Supplier No. Y1002
The kit essentially generates spheroplasts of yeast cells for subsequent analysis. It is optimized for S. cerevisiae and C. albicans. The kit can also be used for any fungus species that is susceptible to yeast lytic enzyme digestion.