Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Yeast PST1 Synthetic Peptide

Catalog No. PEP1456
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1456 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1456 Supplier Invitrogen™ Supplier No. PEP1456
Add to Cart
Add to Cart

Synthetic Peptide

Pst1 Synthetic Peptide, PEP-1456, from Invitrogen.

Protoplast secreted protein 1 (Pst1) is an anchored plasma membrane protein. Pst1 was previously identified as a protein secreted by yeast regenerating protoplasts, which suggests a role in cell wall construction. Pst1 is a protein with 444 amino acids and is attached to yeast cell wall via a glycosylphosphatidylinisotol (GPI) anchor. Pst1 contains 15 potential N-linked glycosylation sites and is heavily glycosylated. It migrates at approximately 200 kDa on SDS-PAGE when produced in wild type S. cerevisiae. Our Pst1 protein was expressed in a genetically manipulated triple-mutant (TM) S. cerevisiae stain, which results in the production of sole Man8GlcNAc2 carbohydrate structures and Pst1 migration at approximately 100 kDa. When produced in the TM yeast, Pst1 can be recognized by several glycan-specific HIV-1 broadly neutralizing antibodies, including 2G12 and recently identified PGT antibodies. Among several heavily N-glycosylated TM yeast glycoproteins, Pst1 shows higher affinity for 2G12 and efficiently inhibits gp120 interactions with 2G12 and DC-SIGN, and it also blocks 2G12-mediated neutralization of HIV-1 pseudoviruses.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 851625
Species Yeast
Purification Method purified
Common Name Yeast PST1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol PST1
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.