Learn More
Biotium Yeast Stains Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF740 5/PK
Catalog No. 5000501226
Encompass_Preferred
Concanavalin A (Con A) is a widely used lectin that selectively binds to a-mannopyranosyl and a-glucopyranosy residues found in the cell wall of yeast and fungi and the cell membrane of mammalian cells Near-infrared CF 740 has Ex/Em at 742/767 nm 1 mg lyophilized solid
Catalog No. 50-005-01226 Supplier Biotium Supplier No. 29136
Concanavalin A (Con A) is a widely used lectin that selectively binds to a-mannopyranosyl and a-glucopyranosy residues found in the cell wall of yeast and fungi and the cell membrane of mammalian cells Near-infrared CF 740 has Ex/Em at 742/767 nm 1 mg lyophilized solid