Learn More
Biotium Yeast Stains Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF790 5/PK
Catalog No. 5000500070
Encompass_Preferred
Concanavalin A (Con A) is a widely used lectin that selectively binds to a-mannopyranosyl and a-glucopyranosy residues found in the cell wall of yeast and fungi and the cell membrane of mammalian cells Near-infrared CF790 has Ex/Em at 783/808 nm 1 mg lyophilized solid
Catalog No. 50-005-00070 Supplier Biotium Supplier No. 29142
Concanavalin A (Con A) is a widely used lectin that selectively binds to a-mannopyranosyl and a-glucopyranosy residues found in the cell wall of yeast and fungi and the cell membrane of mammalian cells Near-infrared CF790 has Ex/Em at 783/808 nm 1 mg lyophilized solid