Learn More
Yeast Synthetic Drop-out Medium Supplements without Leucine and Tryptophan, MilliporeSigma™ Supelco™
Catalog No. 111004447 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 g
Catalog No. Quantity
11-100-4447 20 g
1 options
Catalog No. 11-100-4447 Supplier MilliporeSigma Supelco Supplier No. Y075020G
Add to Cart
Add to Cart
Mixtures of amino acids and other nutrients to be added to Yeast Nitrogen Base Without Amino Acids.
TRUSTED_SUSTAINABILITY
Specifications
Description The supplements contain all of the following components except as indicated:Amino acids: All standard amino acids except for leucine and tryptophan. All the amino acids are present at a concentration of 76 mg/LOther nutrients: Adenine (18 mg/L), inositol (76 mg/L), p-aminobenzoic acid (8 mg/L), uracil (76 mg/L).
Format Poly Bottle
Quantity 20 g