Learn More
Yeast Synthetic Drop-out Medium Supplements without Uracil, MilliporeSigma™ Supelco™
Catalog No. 111004559 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 g
Catalog No. Quantity
11-100-4559 20 g
1 options
Catalog No. 11-100-4559 Supplier MilliporeSigma Supelco Supplier No. Y150120G
Add to Cart
Add to Cart
Mixtures of amino acids and other nutrients to be added to Yeast Nitrogen Base Without Amino Acids.
TRUSTED_SUSTAINABILITY
Specifications
Description The supplements contain all of the following components except as indicated:Amino acids: All standard amino acids are present at a concentration of 76 mg/L except for leucine, which is present (at 380 mg/LOther nutrients: Adenine (18 mg/L), inositol (76 mg/L), p-aminobenzoic acid (8 mg/L))
Format Poly Bottle
Quantity 20 g