Learn More
Bioworld YEAST THIOREDOXIN (RECOMBIN
Supplier:  Bioworld 220607111
Encompass_Preferred
YEAST THIOREDOXIN (RECOMBINANT): Recombinant yeast Thioredoxin (Trx) Source: E.Coli Background: Thioredoxins are a group of ca. 12 kDa redox proteins that mediate numerous cytosolic processes in all cells. Human thioredoxin can be ex...
Catalog No. 50-253-809