Learn More
Biotium Yeast Viability Apoptosis and Viability Assays Dead cell stains Live or Dye 850 870 1/EA
Numéro de catalogue. 5000501500
Encompass_Preferred
Live-or-Dye Fixable Viability Staining Kits are designed for discrimination between live and dead cells during flow cytometry Live-or-Dye 850/870 has emission at 870 nm and can be excited by an 808 laser line 50 assays per kit
Numéro de catalogue. 50-005-01500 Fournisseur Biotium Code fournisseur 32022T
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Live-or-Dye Fixable Viability Staining Kits are designed for discrimination between live and dead cells during flow cytometry Live-or-Dye 850/870 has emission at 870 nm and can be excited by an 808 laser line 50 assays per kit
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.