Learn More
Research Products International Corp YeaStar Genomic DNA Kit w/Zymo-Spin III Columns, Uncapped, 40 Preps
SDP
Catalog No. 501977663
Encompass
The YeaStar Genomic DNA Kit is designed for reliable and efficient isolation of genomic DNA from a broad spectrum of fungal species, including Aspergillus fumigatus, Aspergillus nidulans, Aspergillus nivens var. aureus, Candida albicans, Pichia pastoris, Saccharomyces cerevisiae, Schizosaccharomyces pombe, and any fungi whose cell walls are susceptible to yeast lytic enzyme lysis. The kit is based on highly efficient enzyme lysis and Zymo-Spin column technology. Each standard prep yields about 7 - 20 µg of DNA with a size distribution of 35 - 60 kb. The resulting genomic DNA can be used direct analysis including Southern blotting, PCR, restriction endonuclease digestion, etc.
Simple: Fast spin-column procedure yields pure yeast genomic DNA without using glass beads or phenol.
Catalog No. 50-197-7663 Supplier Research Products International Corp Supplier No. ZD2002
The YeaStar Genomic DNA Kit is designed for reliable and efficient isolation of genomic DNA from a broad spectrum of fungal species, including Aspergillus fumigatus, Aspergillus nidulans, Aspergillus nivens var. aureus, Candida albicans, Pichia pastoris, Saccharomyces cerevisiae, Schizosaccharomyces pombe, and any fungi whose cell walls are susceptible to yeast lytic enzyme lysis. The kit is based on highly efficient enzyme lysis and Zymo-Spin column technology. Each standard prep yields about 7 - 20 µg of DNA with a size distribution of 35 - 60 kb. The resulting genomic DNA can be used direct analysis including Southern blotting, PCR, restriction endonuclease digestion, etc.
Simple: Fast spin-column procedure yields pure yeast genomic DNA without using glass beads or phenol.