Learn More
Zymo Research Corporation YeaStar™ Genomic DNA Kit (40 Preps) w/ Zymo-Spin™ III Columns (Uncapped)[Includes E1006 x 1: R-Zymolyase (1000 Units Lyophilized) w/ Storage Buffer (500 µl)]
SDP
Catalog No. NC9727259
Encompass_Preferred
The YeaStar Genomic DNA Kit™ is designed for reliable and efficient isolation of genomic DNA from a broad spectrum of fungus species, including Aspergills fumigatus, Aspergills nidulans, Aspergills nivens var.aureus, Candida albicans, Pichia pastoris, Saccharomyces cerevisiae, Schizosaccharomyces pombe, and any fungi whose cell walls are susceptible to yeast lytic enzyme lysis. The kit is based on highly efficient enzyme lysis and fast spin column technology. Each standard prep yields about 7-20 g of DNA with a size distribution of 35-60 kb. The resulting genomic DNA can be used directly for all molecular biology analysis such as Southern Blot, PCR, restriction enzyme digestion, etc.
Catalog No. NC9727259 Supplier Zymo Research Corporation Supplier No. D2002
The YeaStar Genomic DNA Kit™ is designed for reliable and efficient isolation of genomic DNA from a broad spectrum of fungus species, including Aspergills fumigatus, Aspergills nidulans, Aspergills nivens var.aureus, Candida albicans, Pichia pastoris, Saccharomyces cerevisiae, Schizosaccharomyces pombe, and any fungi whose cell walls are susceptible to yeast lytic enzyme lysis. The kit is based on highly efficient enzyme lysis and fast spin column technology. Each standard prep yields about 7-20 g of DNA with a size distribution of 35-60 kb. The resulting genomic DNA can be used directly for all molecular biology analysis such as Southern Blot, PCR, restriction enzyme digestion, etc.