Learn More
Zymo Research Corporation YeaStar™ RNA Kit (40 Preps) w/ Zymo-Spin™ IIIC Columns (Capped)[Includes E1004 x 1: Zymolyase (1000 Units Lyophilized) w/ Storage Buffer (500µl)]
SDP
Catalog No. 50444556
Encompass_Preferred
Generally, the kit can be used for the purification of high-quality, total RNA from any fungus that can be lysed by yeast lytic enzyme. The kit facilitates the purification of 10-25 g of total RNA from 1-1.5 ml of cultured cells using innovative Zymo-Spin column technology.
Catalog No. 50-444-556 Supplier Zymo Research Corporation Supplier No. R1002
Generally, the kit can be used for the purification of high-quality, total RNA from any fungus that can be lysed by yeast lytic enzyme. The kit facilitates the purification of 10-25 g of total RNA from 1-1.5 ml of cultured cells using innovative Zymo-Spin column technology.