Learn More
Aviva Systems Biology Corp YEATS4 Antibody - C-terminal region (ARP30117_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30117_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against YEATS4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human YEATS4. Alternate names: 4930573H17Rik, B230215M10Rik, GAS41, NUBI-1, YAF9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-54134