Learn More
Aviva Systems Biology Corp YEATS4 Antibody - middle region (ARP30118_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30118_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against YEATS4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human YEATS4. Alternate names: YAF9, GAS41, NUBI-1, 4930573H17Rik, B230215M10RikApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-54135