Learn More
Bioworld YEB Agrobacterium Growth Medium, 500 g
Catalog No. 50254815
Encompass_Preferred
YEB Agrobacterium Growth MediumOne of the best medium for argrobacterium growth pH: 7.2Composition:Beef Extract Yeast Extract Peptone Sucrose Magnesium Sulfate Use 16.3g/L
Catalog No. 50-254-815 Supplier Bioworld Supplier No. 306270311
YEB Agrobacterium Growth MediumOne of the best medium for argrobacterium growth pH: 7.2Composition:Beef Extract Yeast Extract Peptone Sucrose Magnesium Sulfate Use 16.3g/L