Learn More
Enzo Life Sciences YEEI peptide (biotinylated) (200µg)
SDP
Supplier:  Enzo Life Sciences BMLP3020200
Encompass_Preferred
This biotinylated probe binds to SH2 domains of Src family members including Src, Lck, and Fyn. Purity: ≥97%. Solubility: Soluble in Water. Long Term Storage: -20°C.
Catalog No. 50-201-1334