Learn More
Chemglass Life Sciences PTFE Color-Coded Clamp, 14/35 Standard Taper Joint (Yellow)
SDP
Catalog No. NC2611682
Encompass_Preferred
PTFE color-coded clamp designed to secure 14/35 standard taper glass joints. The PTFE construction provides chemical resistance and minimizes contamination, while the yellow color coding enables rapid size identification in multi-component assemblies.
  • PTFE construction for chemical resistance and low contamination
  • Fits 14/35 standard taper joints
  • Color coded yellow for quick size identification
  • Sold individually
  • Lightweight and corrosion resistant
Catalog No. NC2611682 Supplier Chemglass Life Sciences Supplier No. CG145P14
PTFE color-coded clamp designed to secure 14/35 standard taper glass joints. The PTFE construction provides chemical resistance and minimizes contamination, while the yellow color coding enables rapid size identification in multi-component assemblies.
  • PTFE construction for chemical resistance and low contamination
  • Fits 14/35 standard taper joints
  • Color coded yellow for quick size identification
  • Sold individually
  • Lightweight and corrosion resistant