Learn More
Hach Company Yellow Beaker pH Electrode Cal
Supplier:  Hach Company SM5012
Encompass
Beaker for pH Electrode Calibration-Yellow-30 mL-pk/80Colored and graduated 30 mL plastic beakers It matches buffer color coding to prevent operators from wrong buffer use while calibrating Economic as it ensures optimum amount of buffer is used during calibration steps preventing from excess use of buffers by operators Safe as disposable preventing from contamination
Catalog No. 50-002-07171