Learn More
Leica Biosystems YELLOW CASETTES CS/1000
Catalog No. NC9075654
Encompass_Preferred
IP C Cassettes III are one-piece processing cassettes with a 35 degree writing surface. The cassettes are taped for easy loading into ink printers. Available in eleven colors in quantities of 1000.
Catalog No. NC9075654 Supplier Leica Biosystems Supplier No. 38440203
IP C Cassettes III are one-piece processing cassettes with a 35 degree writing surface. The cassettes are taped for easy loading into ink printers. Available in eleven colors in quantities of 1000.