Learn More
Andwin Scientific Yellow Cryo-Babies 28125 in W x 0.28125 in H.
SDP
Catalog No. NC9760008
Encompass_Preferred
Cryogenic labels formatted for laser printers. The large yellow Tough-Tags labels are conveniently pre-cut to be easily put on microcentrifuge tubes and other laboratory containers. The labels can resist temperatures as low as -20C (-80C when used on virgin polypropylene tubes.) This product can also withstand organic solvents and caustic agents without peeling or falling off. The labels measure 1.28125 in W x 0.28125 in H.
Catalog No. NC9760008 Supplier Andwin Scientific Supplier No. LCRY1700Y
Cryogenic labels formatted for laser printers. The large yellow Tough-Tags labels are conveniently pre-cut to be easily put on microcentrifuge tubes and other laboratory containers. The labels can resist temperatures as low as -20C (-80C when used on virgin polypropylene tubes.) This product can also withstand organic solvents and caustic agents without peeling or falling off. The labels measure 1.28125 in W x 0.28125 in H.