Learn More
Beckman Coulter Beckman Coulter 90 mm Yellow Cup Adapter for Blood Bags
Catalog No. NC2948746
Encompass_Preferred
The Beckman Coulter 90 mm Yellow Cup Adapter (MPN 356856) is a centrifuge cup accessory designed to hold single or double blood bags securely during rotor centrifugation. It is used with compatible swinging-bucket rotors for blood bag processing and separation.
  • Inner diameter 90 mm.
  • Color yellow.
  • Accommodates up to two single blood bags or one double pack.
  • Designed for use in swinging-bucket rotors.
  • Provides secure positioning of blood bags during centrifugation.
Catalog No. NC2948746 Supplier Beckman Coulter Supplier No. 356856
The Beckman Coulter 90 mm Yellow Cup Adapter (MPN 356856) is a centrifuge cup accessory designed to hold single or double blood bags securely during rotor centrifugation. It is used with compatible swinging-bucket rotors for blood bag processing and separation.
  • Inner diameter 90 mm.
  • Color yellow.
  • Accommodates up to two single blood bags or one double pack.
  • Designed for use in swinging-bucket rotors.
  • Provides secure positioning of blood bags during centrifugation.