Learn More
Bioss Yellow Fever virus
Supplier: Bioss BSM72474M
Yellow Fever virus Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide