Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yellow Industrial Flashlights
SCI_ED

Catalog No. S43915IN Shop All Rayovac Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S43915IN Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S43915IN Supplier Rayovac Supplier No. IN2D

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Requires

Both types use two D cells (sold separately).

Tough and durable enough for student use!

With standard lamp or krypton lamp.

Specifications

Battery Life 20 hr.
Battery Type 2 Cell D
Color Yellow
For Use With (Equipment) For toolbox, vehicle
Includes Ring hanger, standard lamp or krypton lamp
Lumen 10 lm
Material Polypropylene
Warranty Lifetime
Product Type Industrial Flashlight
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.