Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hillrom Yellow Isolation Stethsocope

Catalog No. 22355150
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-355-150 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-355-150 Supplier Hillrom Supplier No. WEL 59803
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stethoscope for isolation situations - The chest piece and binaurals are metal, not plastic, for clear transmission of respiratory sounds.

  • For isolation use
  • Yellow color for easy identification
  • Metal chest piece and binaurals

Specifications

Product Type Stethoscope
Color Yellow
For Use With (Application) Isolation Use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.