Learn More
Checkers Yellow Jacket™ 2.25 in. Max Diameter AMS, ADA Male Ramps YJ225-RMP Checkers
Catalog No. 19088404 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Yellow
Catalog No. Color
19-088-404 Yellow
1 options
Catalog No. 19-088-404 Supplier Checkers Supplier No. WSA225RMAHDY
Add to Cart
Add to Cart
Use the 3-Channel Yellow Jacket™ AMS™ Accessibility Female ADA Ramp with The YELLOW JACKET™ 3-CHANNEL AMS™. The YELLOW JACKET™ 3-CHANNEL AMS™ utilizes separate cable protector sections and ramps to create a system that is fully expandable to hold an unlimited number of cables.
  • AMS Ramps Sold Individually
  • AMS Female Ramp = YJ3-225-AMS-F-B
  • AMS Male Ramp - YJ3-225-AMS-M-B
Specifications
Color Yellow
Material HDPE
Product Type Cable Protector Kit