Learn More
Checkers Yellow Jacket™ 3 Channel YJ225 Checkers
Catalog No. 19088402 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Black on Yellow
Catalog No. Color
19-088-402 Black on Yellow
1 options
Catalog No. 19-088-402 Supplier Checkers Supplier No. YJ3225Y/B
Only null left
Add to Cart
Add to Cart
Yellow Jacket™ 3-channel cable protector with hinged lid provides a safer crossing for vehicles and pedestrian traffic while protecting valuable electrical cables and hose lines from damage.
  • Protects cables and hoses up to 2.125 in. outside dia.
  • Dog bone connectors to extend to any desired length
  • Thick lid and base allow for high volume of traffic
  • Hinged lid for easy cable placement
  • Modular interlocking design
  • Ideal for use in industrial, commercial, and public applications
Specifications
Color Black on Yellow
Product Type Cable Protector
Material Polyurethane