Learn More
Checkers Yellow Jacket™ AMS™ Modular Accessibility Ramp System
Catalog No. 17456118 Shop All Justrite Safety Group Products
Change view
Click to view available options
Length (Metric):
1.81 m
Catalog No. Length (Metric)
17-456-118 1.81 m
1 options
Catalog No. 17-456-118 Supplier Checkers Supplier No. WSA225KIT1HS
Add to Cart
Add to Cart
Kit 1, 3-Channel Modular Accessibility Ramp System.
  • Kit 1 - Includes:
    • 2 Ramps
    • 4 Blue Side Ramps
    • 2 AMS™ Center Sections
    • Dog Bone™ Connectors
  • 3-Channel Yellow Jacket™ AMS™ (Advanced Modular System) Accessibility Ramps - Kit 1 features a gradual slope and slip-resistant surface to ensure a safe crossing point for carts, forklifts, vehicles with small wheels and heavy trucks.
  • In addition, each Checkers™ accessibility ramp system is manufactured to meet ADA and DDA compliant specifications
Specifications
Type Cable Tie Assortment Kit
Material Plastic
Color Yellow
Length (Metric) 1.81 m