Learn More
USA Scientific Inc Cryo-Babies, 1.28 inch x 0.5 inch, yellow, on sheets
Catalog No. NC9532771
Encompass
Cryo-Babies, large, yellow labels for 1.5 mL and 2.0 mL tubes, 1.28 inch L x 0.5 inch W, 85/sheet, 1700/box
Catalog No. NC9532771 Supplier USA Scientific Inc Supplier No. 91871706
Cryo-Babies, large, yellow labels for 1.5 mL and 2.0 mL tubes, 1.28 inch L x 0.5 inch W, 85/sheet, 1700/box